Phenol 99,0-100,5%, EMPROVE® ESSENTIAL USP, Ph. Eur., ChP, JP, reinst, SAFC®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Phenylalkohol, Hydroxybenzol, Carbolsäure
1.00201.1000
1.00201.9025
1.00201.1000EA
336
EUR
1.00201.1000
1.00201.9025
Phenol 99,0-100,5%, EMPROVE® ESSENTIAL USP, Ph. Eur., ChP, JP, reinst, SAFC®
Phenol
For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Formel:
C₆H₅OH Molecular Weight: 94.11 g/mol Siedepunkt: 181,7 Schmelzpunkt: 40,8 Dichte: 1.0722 g/cm³ (20 °C) Flammpunkt: 81 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002143 CAS-Nummer: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1 II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Spezifikation Testergebnisse
Assay (bromatometric) | 99.0 - 100.5 % |
Assay (bromatometric, calc. on anhydrous substance) | 99.0 - 100.5 % |
Identity (IR-spectrum) | passes test |
Identity (wet chemistry) | passes test |
Appearance | colourless or faintly pink or faintly yellowish crystals or crystalline masses, deliquescent |
Appearance of solution (67 g/l, water) | clear and not more intensely coloured than reference solution B6 |
Acidic substances | passes test |
Solidification temperature | ≥ 39.5 °C |
Fe (Iron) | ≤ 1 ppm |
Benzene (HS-GC) | ≤ 2 ppm |
Methanol (HS-GC) | ≤ 3000 ppm |
Acetone (HS-GC) | ≤ 2 ppm |
Anisole | ≤ 10 ppm |
Cumene (HS-GC) | ≤ 70 ppm |
Methylisobutylketone (HS-GC) | ≤ 2 ppm |
Other residual solvents (ICH (Q3C)) | excluded by manufacturing process |
Residue on evaporation (105 °C, 1 h) | ≤ 0.05 % |
Residue on evaporation (105 °C) | ≤ 0.05 % |
Water (according to Karl Fischer) | ≤ 0.5 % |
Corresponds to Ph. Eur., ChP, JP, USP |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...