Salzsäure 25%, EMSURE® zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ300
1.00316.1000
1.00316.1011
1.00316.2500
1.00316.2511
1.00316.9025
1.00316.9950
1.00316.1000EA
35.1
EUR
1.00316.1000
1.00316.1011
1.00316.2500
1.00316.2511
1.00316.9025
1.00316.9950
Salzsäure 25%, EMSURE® zur Analyse, Supelco®
Salzsäure
For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues
Formel:
HCl Siedepunkt: 107 °C (1013 hPa) Schmelzpunkt: –52 °C Dichte: 1,12 g/cm3 (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Spezifikation Testergebnisse
Assay (Alkalimetric) | ≥25.0% |
Color | ≤10 Hazen |
Bromide (Br) | ≤50 ppm |
Free Chlorine (Cl) | ≤0.5 ppm |
Sulfate (SO₄) | ≤1 ppm |
Sulfite (SO₃) | ≤1 ppm |
Al (Aluminium) | ≤0.050 ppm |
As (Arsenic) | ≤0.01 ppm |
Ba (Barium) | ≤0.020 ppm |
Be (Beryllium) | ≤0.020 ppm |
Bi (Bismuth) | ≤0.100 ppm |
Ca (Calcium) | ≤0.500 ppm |
Cd (Cadmium) | ≤0.010 ppm |
Co (Cobalt) | ≤0.010 ppm |
Cr (Chromium) | ≤0.020 ppm |
Cu (Copper) | ≤0.020 ppm |
Fe (Iron) | ≤0.200 ppm |
Ge (Germanium) | ≤0.050 ppm |
K (Potassium) | ≤0.100 ppm |
Li (Lithium) | ≤0.010 ppm |
Mg (Magnesium) | ≤0.100 ppm |
Mn (Manganese) | ≤0.010 ppm |
Mo (Molybdenum) | ≤0.020 ppm |
NH₄ (Ammonium) | ≤1 ppm |
Na (Sodium) | ≤0.500 ppm |
Ni (Nickel) | ≤0.020 ppm |
Pb (Lead) | ≤0.020 ppm |
Sr (Strontium) | ≤0.010 ppm |
Ti (Titanium) | ≤0.100 ppm |
Tl (Thallium) | ≤0.050 ppm |
V (Vanadium) | ≤0.010 ppm |
Zn (Zinc) | ≤0.050 ppm |
Zr (Zirconium) | ≤0.100 ppm |
Residue on Ignition (as SO₄) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...