Silver plating stain kit acc. to von Kossa zur Kalziumbestimmung, Färbemittel für Histologie, Sigma-Aldrich®

Lieferant: Merck
Achtung

1.00362.0001
1.00362.0001EA 187 EUR
1.00362.0001
Silver plating stain kit acc. to von Kossa zur Kalziumbestimmung, Färbemittel für Histologie, Sigma-Aldrich®
Silver plating stain kit

For detection of calcium deposits in paraffin sections


Tissue calcification can interfere with normal metabolic processes and occurs in various tissues, like bone marrow, mamma and also in tumors. The Silver Staining Kit acc. to von Kossa is used to visualize calcium deposits in histological tissue specimens. The Kit contains two ready-to-use reagents, 100 ml of a Silver nitrate solution and 100 ml of a Sodium thiosulfate solution. The kit is sufficient for up to 300 slides. As counterstain, the nuclei could be visualized with 0.1% Nuclear fast red-aluminum sulfate solution. The product is registered as IVD and CE product and can be used in diagnostics and laboratory accreditation.

Lagertemperatur: Raumtemperatur
UN: 3316
ADR: 9,II
IVDLogo

Bestellen

Spezifikation Testergebnisse

Suitability for microscopy passes test
Calcium brown to black
Nuclei red
Collagen red
Background red


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR