Salpetersäure 65%, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.00443.1000 1.00443.2500 1.00443.9025
1.00443.1000EA 38.4 EUR
1.00443.1000 1.00443.2500 1.00443.9025
Salpetersäure 65%, EMPLURA®, Supelco®
Salpetersäure
Formel: HNO₃
Siedepunkt: 120,5 °C (1013 hPa)
Schmelzpunkt: –42 °C
Dichte: 1,4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric) 64.3 - 66.4 %
Identity passes test
Appearance of solution passes test
Chloride (Cl) ≤ 0.0003 %
Iodate, Bromate passes test
Nitrogen oxides (as N₂O₃) ≤ 0.003 %
Sulphate (SO₄) ≤ 0.001 %
Heavy metals (as Pb) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Ca (Calcium) ≤ 0.001 %
Fe (Iron) ≤ 0.0004 %
NH₄ (Ammonium) ≤ 0.001 %
Residual solvents (ICH Q3C) ...excluded by production process
Evaporation residue ≤ 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR