Masson-Goldner staining kit für die Histologie, Sigma-Aldrich®
Lieferant: Merck
Gefahr
1.00485.0001
1.00485.0001EA 497 EUR
1.00485.0001
Masson-Goldner staining kit für die Histologie, Sigma-Aldrich®
Masson-Goldner staining kit
Visualisation of connective tissue
Connective tissue can be selectively visualised by using a combination of three different staining solutions: Azophloxine and tungstophosphoric acid Orange G solutions stain components such as muscle, cytoplasm and erythrocytes, and Light green SF solution then is counterstained. These solutions are used in the Masson-Goldner trichrome staining technique which has the benefit that it can be used on formalin-fixed material. Our Masson-Goldner staining kit contains the three ready to use staining solutions that can also be applied subsequently to staining nuclei with Wiegert's iron haematoxylin. As result of a staining according to our protocol, the nuclei will be stained in dark brown to black, cytoplasm and muscles will appear brick red, connective tissue and acidic mucus substances will appear green and erythrocytes will be bright orange.The package is sufficient for 400 to 500 applications. The product is registered as a IVD and CE product and can be used in diagnostics and laboratory accreditations.
UN: 2790
ADR: 8,III
IVDLogo
Bestellen
Spezifikation Testergebnisse
Suitability for microscopy (tissue section) Passes test
Nuclei Dark brown to black
Cytoplasm Brick red
Musculature (muscles) Bright red
Connective tissue Green
Acid mucosubstances Green
Erythrocytes Bright orange