Parteck® Magnesiumstearat, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, FCC vegetable grade, SAFC®
Lieferant: Merck
|
Achtung
|
Synonyme:
Stearinsäure Magnesiumsalz
1.00663.9020
1.00663.0250
1.00663.9020EA
0
EUR
1.00663.9020
1.00663.0250
Parteck® Magnesiumstearat, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, FCC vegetable grade, SAFC®
Magnesiumstearat
Residues of metal catalysts or metal reagents acc. to EMEA/CHMP/SWP/4446/2000 are not likely to be present.
Corresponds to Ph Eur, BP, JP, NF, FCC
Formel:
C₃₆H₇₀MgO₄ Molecular Weight: 591.25 g/mol Schmelzpunkt: 130…140 °C Dichte: 1.03 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036391 CAS-Nummer: 557-04-0 EINECS: 209-150-3 Merck Index: 13,05714 |
Spezifikation Testergebnisse
Assay (Mg; calculated on dried substance) | 4.0 - 5.0 % |
Identity | passes test |
Palmitic- stearic acid components of fatty acids | ≥ 95 % |
Stearic acid component of fatty acids | 60 - 70 % |
Acidity or alkalinity | passes test |
Chloride (Cl) | ≤ 0.10 % |
Sulphate (SO₄) | ≤ 0.3 % |
Heavy metals (as Pb) | ≤ 0.0020 % |
Cd (Cadmium) | ≤ 0.0001 % |
Ni (Nickel) | ≤ 0.00050 % |
Pb (Lead) | ≤ 0.0005 % |
Residual solvents (Ph.Eur./USP/ICH) | excluded by the production process |
Acid value | 195 - 210 |
Loss on Drying (105°C) | ≤ 4.0 % |
Particle Size (d10) | ≥ 1 µm |
Particle Size (d50) | about 5 |
Particle Size (d90) | ≤ 20 µm |
Specific surface area (BET) | 5 - 12 m²/g |
Microbiological purity (Total aerobic microbial count (TAMC)) | ≤ 1E+03 CFU/g |
Microbiological purity (Total yeast and mould count (TYMC)) | ≤ 1E+02 CFU/g |
Bile tolerant Gram negative Bacteria (absent in 1 g) | passes test |
Escherichia coli (absent in 1 g) | passes test |
Pseudomonas aeruginosa (absent in 1 g) | passes test |
Staphylococcus aureus (absent in 1 g) | passes test |
Candida albicans (absent in 1 g) | passes test |
Salmonella ssp (absent in 10 g) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...