Diisopropylether, EMSURE® Reag. Ph. Eur., ACS zur Analyse, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Isopropyl ether
M-Clarity™ Qualitätsstufe = MQ300
1.00867.1000
1.00867.2500
1.00867.6010
1.00867.6190
1.00867.4000
1.00867.1000EA
63.1
EUR
1.00867.1000
1.00867.2500
1.00867.6010
1.00867.6190
1.00867.4000
Diisopropylether, EMSURE® Reag. Ph. Eur., ACS zur Analyse, Supelco®
Diisopropylether
EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Formel:
(CH₃)₂CHOCH(CH₃)₂ Siedepunkt: 68,5 °C (1013 hPa) Schmelzpunkt: –86 °C Dichte: 0,726 g/cm³ (20 °C) Flammpunkt: –28 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008880 CAS-Nummer: 108-20-3 EINECS: 203-560-6 UN: 1159 ADR: 3,II Merck Index: 13,05232 |
|
Spezifikation Testergebnisse
Solubility | 12 g/l (20 °C) |
Melting Point | -86 °C |
Molar Mass | 102.18 g/mol |
Boiling Point | 67 - 70 °C (1013 hPa) |
Vapor Pressure | 175 hPa (20 °C) |
Flash Point | -28 °C |
Refractive Index | 1.3679 (20 °C, 589 nm) |
Explosion Limit | 1.0 - 21 %(V) |
Density | 0.72 g/cm3 (20 °C) |
pH | (H2O, 20 °C) neutral |
Ignition Point | 405 °C |
Evaporation number | 1.6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...