Ethylacetat, LiChrosolv® für die HPLC, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Essigester, Essigsäureethylester, EtOAc

M-Clarity™ Qualitätsstufe = MQ100

1.00868.1000 1.00868.2500 1.00868.9185
1.00868.1000EA 64.04 EUR
1.00868.1000 1.00868.2500 1.00868.9185
Ethylacetat, LiChrosolv® für die HPLC, Supelco®
Ethylacetat

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formel: CH₃COOC₂H₅
Molecular Weight: 88.11 g/mol
Siedepunkt: 77.1
Schmelzpunkt: –83 °C
Dichte: 0.902 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 85.3 g/l (20 °C)
Melting Point -83 °C
Molar Mass 88.11 g/mol
Boiling Point 77 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point -4 °C
Refractive Index 1.37
Explosion Limit 2.1 - 11.5 %(V)
Density 0.90 g/cm3 (20 °C)
pH
Ignition Point 460 °C
Viscosity kinematic
Synonyms Ethyl ethanoate, Acetic ester, Ethyl ester
Hill Formula C4H8O2
Chemical Formula CH3COOC2H5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR