Saccharose, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, ChP excipient_suitable Endotoxins_low, SAFC®

Lieferant: Merck

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

MQ500

1.00892.9050 1.00892.5000 1.00892.9029
1.00892.9050EA 1620 EUR
1.00892.9050 1.00892.5000 1.00892.9029
Saccharose, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, ChP excipient_suitable Endotoxins_low, SAFC®
D(+)-Saccharose
Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 185.5 °C
Dichte: 1.59 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity (wet chemistry) passes test
Identity (IR) passes test
Appearance of solution (50 %; water) passes test
Color of solution passes test
Color number ≤ 45
Spec. rotation (α 20/D; 26 %; water) 66.3 - 67.0 °
Spec. rotation (α 20/D; 10 %; water) 66.3 - 67.0 °
Electrical conductivity ≤ 35 µS/cm
Sulfite (as SO₂) ≤ 10 ppm
Sulfate (SO₄) ≤ 0.05 %
Heavy metals (as Pb) ≤ 0.0005 %
Ca (Calcium) ≤ 0.05 %
Dextrins passes test
Reducing sugars passes test
Reducing sugars (ChP) ≤ 0.10 %
Residual solvents (ICH Q3C) excluded by production process
Loss on Drying (105 °C) ≤ 0.1 %
Sulfated ash ≤ 0.1 %
Endotoxins < 2
Microbiological purity (Total aerobic microbial count (TAMC)) ≤ 500 CFU/g
Microbiological purity (Total yeast and mould count (TYMC)) ≤ 100 CFU/g
Bile tolerant Gram negative Bacteria (absent in 1 g) passes test
Escherichia coli (absent in 1 g) passes test
Pseudomonas aeruginosa (absent in 1 g) passes test
Staphylococcus aureus (absent in 1 g) passes test
Candida albicans (absent in 1 g) passes test
Salmonella ssp (absent in 10 g) passes test
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, BP, ChP, JP, NF

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR