Diethylether stabilisiert, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Gefahr


Stabilisierung: Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol 5 - 10 ppm
M-Clarity™ Qualitätsstufe = MQ300

1.00921.1000 1.00921.6010 1.00921.6025 1.00921.2500 1.00921.6190
1.00921.1000EA 46.79 EUR
1.00921.1000 1.00921.6010 1.00921.6025 1.00921.2500 1.00921.6190
Diethylether stabilisiert, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Diethylether

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.


EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: (CH₃CH₂)₂O
Siedepunkt: 35 °C (1013 hPa)
Schmelzpunkt: –123 °C
Dichte: 0,7135 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011646
CAS-Nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (GC) ≥ 99.7 %
Identity (IR) conforms
Appearance clear
Color ≤ 10 Hazen
Titrable acid ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Density (d 20 °C/20 °C) 0.713 - 0.715
Boiling point 34 - 35 °C
Chloride (Cl) ≤ 0.00003 %
Sulfate (SO₄) ≤ 0.00003 %
Acetone (GC) ≤ 0.005 %
Ethanol (GC) ≤ 0.02 %
Methanol (GC) ≤ 0.02 %
Peroxide (as H₂O₂) ≤ 0.000015 %
Carbonyl compounds (as HCHO) ≤ 0.001 %
Carbonyl compounds (as CO) ≤ 0.001 %
Matter discolored by H₂SO₄ ≤ 10 Hazen
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %
Evaporation residue ≤ 0.0005 %
Water ≤ 0.03 %
The product is stabilized with 5 - 10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT). -


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR