Benzylalkohol 98,0-100,5 GC_ASSAY_METHOD, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP excipient_suitable, SAFC®

Lieferant: Merck
Achtung

Synonyme: Phenylcarbinol, (Hydroxymethyl)benzol, Phenylmethanol

1.00981.2500 1.00981.6025 1.00981.6190
1.00981.2500EA 329 EUR
1.00981.2500 1.00981.6025 1.00981.6190
Benzylalkohol 98,0-100,5 GC_ASSAY_METHOD, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP excipient_suitable, SAFC®
Benzylalkohol

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Conforms to Ph Eur, BP, JP, NF

Formel: C₆H₅CH₂OH
Molecular Weight: 108.14 g/mol
Siedepunkt: 205
Schmelzpunkt: –15.3 °C
Dichte: 1.05 g/cm³ (20 °C)
Flammpunkt: 101
Lagertemperatur: Raumtemperatur
MDL: MFCD00004599
CAS-Nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) 98.0 - 100.5%
Identity (IR) Conforms
Acidity Conforms
Refractive Index (n 20/D) 1.538 - 1.541
Density (d 20 °C/20 °C) 1.043 - 1.049
Peroxide Value ≤5.0
Related Substances (GC) (Benzaldehyde (Impurity A)) ≤0.15%
Related Substances (GC) (Cyclohexylmethanol (Impurity B)) ≤0.10%
Related Substances (GC) (Sum of Peaks with a Relative Retention Less than that of Benzyl Alcohol) ≤0.04%
Related Substances (GC) (Sum of Peaks with a Relative Retention Greater than that of Benzyl Alcohol) ≤0.3%
Related Substances (GC) (Disregard Limit) ≤0.0001%
Benzene (GC) ≤0.0002%
Chlorobenzene (GC) ≤0.01%
Toluene (GC) ≤0.01%
Other Residual Solvents (ICH Q3C) Excluded by Manufacturing Process
Evaporation Residue ≤0.050%
Water ≤0.10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR