1-Propanol, EMSURE® ACS, Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.00997.1000 1.00997.2500 1.00997.6025 1.00997.4000
1.00997.1000EA 116 EUR
1.00997.1000 1.00997.2500 1.00997.6025 1.00997.4000
1-Propanol, EMSURE® ACS, Reag. Ph. Eur. zur Analyse, Supelco®
1-Propanol

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: H₃CCH₂CH₂OH
Siedepunkt: 97 °C (1013 hPa)
Schmelzpunkt: –127 °C
Dichte: 0,8053 g/cm³ (20 °C)
Flammpunkt: 15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002941
CAS-Nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) miscible in all proportions
Melting Point -127 °C
Molar Mass 60.1 g/mol
Boiling Point 96.5 - 98 °C (1013 hPa)
Vapor Pressure 19 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 2.1 - 19.2 %(V)
Density 0.80 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 360 °C
Evaporation number 17


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR