Stärke, löslich, extra_pure pH 6.0 - 7.5 (20 g/l, H₂O, 25 °C)., SAFC®

Lieferant: Merck

Synonyme: Amylodextrin, Vitex® Iodindikator

1.01253.1000 1.01253.9025
1.01253.1000EA 1930 EUR
1.01253.1000 1.01253.9025
Stärke, löslich, extra_pure pH 6.0 - 7.5 (20 g/l, H₂O, 25 °C)., SAFC®
Stärke, löslich
Formel: (C₆H₁₀O₅)n
Molecular Weight: 162.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082026
CAS-Nummer: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Bestellen

Spezifikation Testergebnisse

Appearance of solution (color) colourless
Appearance of solution (clearness) almost clear to slightly opalescent
Sensitivity with iodine solution passes test
Reducing substances passes test
Microbiological test passes test
Colony count (aerobic bacteria) ≤ 5000 CFU/g
Colony count (Yeasts and moulds) ≤ 500 CFU/g
Salmonella species absent in 10 g
Staphylococcus aureus absent in 1 g
Pseudomonas aeruginosa absent in 1 g
E.coli absent in 1 g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR