Wright's Färbung -Lösung
Lieferant: Merck
|
Gefahr
|
Synonyme:
Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
Used to stain blood, bone marrow smears, and clinical cytological specimens
The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.
Dichte:
0.80 g/cm3 (20 °C) Flammpunkt: 9.7 °C |
MDL:
MFCD00082143 CAS-Nummer: 68988-92-1 UN: 1230 ADR: 3 (6.1) II |
![]() |
Spezifikation Testergebnisse
Suitability for microscopy (Blood smear) | passes test |
Erythrocytes | pink to brownish |
Nuclei | blue-violet |
Eosinophilic granules | red to red-brown |
Neutrophilic granules | light violet |
Lymphocyte cytoplasm | Blue |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...