Silberdiethyldithiocarbamat ≥99%, EMSURE® Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Achtung

Synonyme: N,N-Diethyldithiocarbamidsäure Silbersalz, (Diethyldithiocarbamato-S,S')silber, Diethyldithiocarbamidsäure Silbersalz, DETC

M-Clarity™ Qualitätsstufe = MQ100

1.01515.0005
1.01515.0005EA 125 EUR
1.01515.0005
Silberdiethyldithiocarbamat ≥99%, EMSURE® Reag. Ph. Eur. zur Analyse, Supelco®
Silberdiethyldithiocarbamat
Formel: (C₂H₅)₂NCSSAg
Molecular Weight: 256,14 g/mol
Schmelzpunkt: 178…183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004929
CAS-Nummer: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Purity (DSC (differential scanning calorimetry)) ≥ 99 %
Identity (IR-spectrum) passes test
Appearance passes test
Appearance (description) passes test
pH-value (aqueous suspension) 5.0 - 6.5
Nitrate (NO₃) ≤ 0.01 %
Loss on Drying (105 °C) ≤ 0.5 %
Sensitivity (for determination of arsenic) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR