Gram-color Färbekit modifiziert zur Gramfärbung phenolfrei, Sigma-Aldrich®

Lieferant: Merck
Gefahr

1.01603.0001
1.01603.0001EA 309 EUR
1.01603.0001
Gram-color Färbekit modifiziert zur Gramfärbung phenolfrei, Sigma-Aldrich®
Gram color staining kit

Phenol-free staining kit for the Gram staining method


Gram-color modified, phenol-free is a staining set for staining microorganisms. Aniline dyes in the cell tissue of microorganisms form a red dye-iodine complex when exposed to iodine. Sodium hydrogen carbonate enhances the formation of this complex further still. In gram-positive microorganisms the dye-iodine complex cannot be subsequently dissolved from the cells by decolorizing agents such as alcohol or acetone. The cell remains dark blue in color. In gram-negative microorganisms the dye-iodine complex is dissolved and the cell turns red as a result of counterstaining. Gram-color modified, phenol-free is an IVD / CE product. The package is sufficient for 65 - 70 applications.

Lagertemperatur: Raumtemperatur
UN: 1993
ADR: 3,II
IVDLogo

Bestellen

Spezifikation Testergebnisse

Suitability for microscopy passes test
gram-positve microorganisms dark blue
gram-negative microorganisms red


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR