Agar, powder_fine, EMPROVE® ESSENTIAL BP, Ph. Eur., extra_pure, SAFC®

Lieferant: Merck

Synonyme: Agar Agar

1.01615.1000 1.01615.9025
1.01615.1000EA 364 EUR
1.01615.1000 1.01615.9025
Agar, powder_fine, EMPROVE® ESSENTIAL BP, Ph. Eur., extra_pure, SAFC®
Agar
Schmelzpunkt: 90
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR