Kongorot Färbungskit
Lieferant: Merck
|
Gefahr
|
Kit for the detection of amyloid acc. to Highman
The detection of amyloid is relevant for a large group of diseases, such as Alzheimer’s disease, Creutzfeld-Jakob’s disease and bovine spongiform encephalopathy. The Congo red staining is routinely used to visualise amyloid structures with the method acc. to Highman - based on the formation of hydrogen bridge bonds of the reagent with the carbohydrate component of the substrate.
The staining kit contains ready to use congo red solution and KOH differentiation solution. The kit can be used for clinical diagnostics and in laboratory accreditation processes, as it is registered as IVD and CE product.The kit is sufficient for up to 50 applications.
Formel:
C₃₂H₂₂N₆Na₂O₆S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004028 CAS-Nummer: 573-58-0 UN: 3316 ADR: 9,II |
![]() |
Spezifikation Testergebnisse
Suitability for microscopy | passes test |
Amyloid | pink to red; in polarized ligt green metachromasis |
Nuclei | dark blue |
Connective tissue | light red |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...