Alcianblau -Lösung Verwendung in der nuklearen Färbung, für die Histologie, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Alcianblau 8 GX
Used to stain acidic mucosubstances
The combination of PAS staining with Alcian blue allows (at pH 2,5) the additional identification of acidic mucosubstances (glycosaminoglycans). PAS staining is used for paraffin sections and cell smears, and upon use of the ready to use Alcian blue staining solution, nuclei will appear blue, acidic mucosubstances light blue and polysaccharides, as well as neutral mucopolysaccharides, will be stained purple. Additionally, the Alcian blue staining solution (at pH 2,5) allows the identification of acidic mucosubstances without differentiation of carboxyl and sulphate groups, upon which the nuclei are counter-stained with nuclear fast red. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.
Formel:
C₅₆H₆₈Cl₄CuN₁₆S₄ Dichte: 1,006 g/cm3 (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010720 CAS-Nummer: 33864-99-2 |
![]() |
Spezifikation Testergebnisse
Suitability for microscopy | passes test |
neutral mucopolysaccharides | purple |
polysaccharides | purple |
PAS negative acidic mucosubstances | light blue |
Nuclei | Blue |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...