Bariumsulfat für Referenzweiß nach DIN 5033 (Blanc fixe), Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.01748.0250
1.01748.0250EA 794 EUR
1.01748.0250
Bariumsulfat für Referenzweiß nach DIN 5033 (Blanc fixe), Supelco®
Barium sulphate

Certificate of Guarantee: PTB-Test Official number
Spectral radiance factor is calculated from measurement results
Tristimulus values X,Y,Z and reflection factor R457 are calculated from spectral radiance factor β(λ)d/0

Formel: BaSO₄
Molecular Weight: 233,39 g/mol
Siedepunkt: 1600 °C (1013 hPa)
Schmelzpunkt: 1345 °C
Dichte: 4,5 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003455
CAS-Nummer: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Bestellen

Spezifikation Testergebnisse

Spectral radiance factor β(λ)d/₀ (350 nm)
Spectral radiance factor β(λ)d/₀ (360 nm)
Spectral radiance factor β(λ)d/₀ (370 nm)
Spectral radiance factor β(λ)d/₀ (380 nm)
Spectral radiance factor β(λ)d/₀ (390 nm)
Spectral radiance factor β(λ)d/₀ (400 nm)
Spectral radiance factor β(λ)d/₀ (420 nm)
Spectral radiance factor β(λ)d/₀ (450 nm)
Spectral radiance factor β(λ)d/₀ (500 nm)
Spectral radiance factor β(λ)d/₀ (600 nm)
Spectral radiance factor β(λ)d/₀ (700 nm)
Spectral radiance factor β(λ)d/₀ (800 nm)
Spectral radiance factor β(λ)₄₅/₀ (360 nm)
Spectral radiance factor β(λ)₄₅/₀ (370 nm)
Spectral radiance factor β(λ)₄₅/₀ (380 nm)
Spectral radiance factor β(λ)₄₅/₀ (390 nm)
Spectral radiance factor β(λ)₄₅/₀ (400 nm)
Spectral radiance factor β(λ)₄₅/₀ (420 nm)
Spectral radiance factor β(λ)₄₅/₀ (450 nm)
Spectral radiance factor β(λ)₄₅/₀ (500 nm)
Spectral radiance factor β(λ)₄₅/₀ (600 nm)
Spectral radiance factor β(λ)₄₅/₀ (700 nm)
Spectral radiance factor β(λ)₄₅/₀ (800 nm)
Reflection factor R₄₅₇
Standard illuminant A 2° CIE 1931 (X)
Standard illuminant A 2° CIE 1931 (Y)
Standard illuminant A 2° CIE 1931 (Z)
Standard illuminant D65 2° CIE 1931 (X)
Standard illuminant D65 2° CIE 1931 (Y)
Standard illuminant D65 2° CIE 1931 (Z)
Standard illuminant C 2 ° CIE 1931 (X)
Standard illuminant C 2 ° CIE 1931 (Y)
Standard illuminant C 2 ° CIE 1931 (Z)
Standard illuminant A 10° CIE 1964 (X)
Standard illuminant A 10° CIE 1964 (Y)
Standard illuminant A 10° CIE 1964 (Z)
Standard illuminant D65 10 ° CIE 1964 (X)
Standard illuminant D65 10 ° CIE 1964 (Y)
Standard illuminant D65 10 ° CIE 1964 (Z)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR