Benzylbenzoat 99,0-100,5% (durch Alkalimetrie) Ph. Eur., BP, USP, SAFC®

Lieferant: Merck
Achtung

Synonyme: Benzoesäurebenzylester

1.01806.6025
1.01806.6025EA 0 EUR
1.01806.6025
Benzylbenzoat 99,0-100,5% (durch Alkalimetrie) Ph. Eur., BP, USP, SAFC®
Benzylbenzoat

Residues of metal catalysts or metal reagents acc. to EMEA/C HMP/SWP/4446/2000 are not likely to be present. Corresponds to Ph Eur, BP, USP.

Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Siedepunkt: 324 °C (1013 hPa)
Schmelzpunkt: 21 °C
Dichte: 1,12 g/cm³ (20 °C)
Flammpunkt: 158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003075
CAS-Nummer: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Assay (by alcalimetric following saponification) 99.0 - 100.5 %
Identity (IR-spectrum) passes test
Acidity passes test
Refractive index (n 20/D) 1.568 - 1.570
Density (Ph Eur, d 20/20) 1.118 - 1.122
Density (USP, d 25/25) 1.116 - 1.120
Solidification point ≥ 18 °C
Al (Aluminium) ≤ 1 ppm
Benzaldehyde ≤ 0.05 %
Benzene (HS-GC) ≤ 2 ppm
Methanol (HS-GC) ≤ 3000 ppm
Toluene (HS-GC) ≤ 890 ppm
Other residual solvents (ICH Q3C) excluded by production process
Sulfated ash ≤ 0.05 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1 to 3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated. ≤ 0,05 %
Corresponds to Ph. Eur., BP, USP ≤ 2 ppm
Other Residual Solvents (ICH Q3C) Excluded by Production Process
Sulphated Ash ≤ 0,05 

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR