tert-Butylmethylether, LiChrosolv® für die HPLC, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.01845.1000 1.01845.2500 1.01845.9030 1.01845.9185
1.01845.1000EA 146 EUR
1.01845.1000 1.01845.2500 1.01845.9030 1.01845.9185
tert-Butylmethylether, LiChrosolv® für die HPLC, Supelco®
tert-Butylmethylether

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formel: (CH₃)₃COCH₃
Siedepunkt: 55,2 °C (1013 hPa)
Dichte: 0,7405 g/cm³ (20 °C)
Flammpunkt: –28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008812
CAS-Nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR