Bicin, Millipore®
Lieferant: Merck
Synonyme:
N,N-Bis(2-hydroxyethyl)glycin
M-Clarity™ Qualitätsstufe = MQ100
1.01910.9010
1.01910.9010EA
4130
EUR
1.01910.9010
Bicin, Millipore®
Bicin
Formel:
C₆H₁₃NO₄ Schmelzpunkt: 185…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004295 CAS-Nummer: 150-25-4 EINECS: 205-755-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...