1-Butanol, EMSURE® ISO, ACS, Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Gefahr

Synonyme: 1-Butylalkohol, 1-Hydroxybutan, n-Butanol, Propylcarbinol

M-Clarity™ Qualitätsstufe = MQ300

1.01990.1000 1.01990.2500
1.01990.1000EA 59.39 EUR
1.01990.1000 1.01990.2500
1-Butanol, EMSURE® ISO, ACS, Reag. Ph. Eur. zur Analyse, Supelco®
1-Butanol

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: H₃C(CH₂)₂CH₂OH
Siedepunkt: 117,6 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0,812 g/cm³ (20 °C)
Flammpunkt: 30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002964
CAS-Nummer: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 77 g/l (20 °C)
Melting Point -89 °C
Molar Mass 74.12 g/mol
Boiling Point 116 - 118 °C (1013 hPa)
Vapor Pressure 6.7 hPa (20 °C)
Flash Point 34 °C
Refractive Index 1.3993 (20 °C, 589 nm)
Explosion Limit 1.4 - 11.3 %(V)
Density 0.81 g/cm3 (20 °C)
pH 7 (70 g/l, H2O, 20 °C)
Ignition Point 340 °C
Evaporation number 33


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR