tri-Calciumdicitrat Tetrahydrat, EMPROVE® ESSENTIAL DAC, USP, FCC, E333, SAFC®

Lieferant: Merck

Synonyme: Calciumcitrat tribasisch Tetrahydrat, di-Citronensäure Tricalciumsalz Tetrahydrat

1.02092.9025
1.02092.9025EA 0 EUR
1.02092.9025
tri-Calciumdicitrat Tetrahydrat, EMPROVE® ESSENTIAL DAC, USP, FCC, E333, SAFC®
tri-Calciumdicitrat Tetrahydrat
Formel: C₁₂H₁₀Ca₃O₁₄·4H₂O
Molecular Weight: 570.5 g/mol
MDL: MFCD00150786
CAS-Nummer: 5785-44-4
EINECS: 212-391-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR