α-Cyclodextrin, Millipore®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

1.02126.1000
1.02126.1000EA 0 EUR
1.02126.1000
α-Cyclodextrin, Millipore®
α-Cyclodextrin
Formel: C₃₆H₆₀O₃₀
Molecular Weight: 972,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078207
CAS-Nummer: 10016-20-3
EINECS: 233-007-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR