Kohlenstoff (aus Holz), Pulver, Millipore®
Lieferant: Merck
Synonyme:
Charcoal
M-Clarity™ Qualitätsstufe = MQ200
1.02204.9025
1.02204.9025EA
624
EUR
1.02204.9025
Kohlenstoff (aus Holz), Pulver, Millipore®
Carbon
Formel:
C Molecular Weight: 12,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00133992 CAS-Nummer: 7440-44-0 EINECS: 231-153-3 UN: 1361 ADR: 4.2,III |
Spezifikation Testergebnisse
Residue of ignition (600 °C) | ≤7% |
Particle size <0.15 mm | ~99% |
Carbonization | Conforms |
Bulk Density | 410 kg/m3 |
Hill Formula | C |
Chemical Formula | C |
Melting point | 3550 °C |
Bulk density | 410 kg/m3 |
Residue on ignition | ≤7 % |
Particle size | about 99 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...