Chinaldinrot, Supelco®

Lieferant: Merck

Synonyme: 2-(2-(4-Dimethylamino)styryl)-1-ethylchinoliniumiodid

M-Clarity™ Qualitätsstufe = MQ200

1.02282.0005
1.02282.0005EA 204 EUR
1.02282.0005
Chinaldinrot, Supelco®
Chinaldinrot
Formel: C₂₁H₂₃IN₂
Molecular Weight: 430,33 g/mol
Schmelzpunkt: 240 °C
MDL: MFCD00011968
CAS-Nummer: 117-92-0
EINECS: 204-221-5
Merck Index: 12,08230

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) practically insoluble
Melting Point 240 °C (decomposition)
Molar Mass 430.33 g/mol
Bulk Density 290 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR