Calcein Indikator, für die Metalltitration, Supelco®
Lieferant: Merck
Synonyme:
Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
M-Clarity™ Qualitätsstufe = MQ200
1.02315.0005
1.02315.0005EA
240
EUR
1.02315.0005
Calcein Indikator, für die Metalltitration, Supelco®
Calcein
Formel:
C₃₀H₂₆N₂O₁₃ |
MDL:
MFCD00005049 CAS-Nummer: 1461-15-0 EINECS: 215-957-1 |
Spezifikation Testergebnisse
Appearance of solution | conforms |
Loss on drying (110 °C) | ≤ 7.0 % |
Suitability as indicator (for metal titration) | conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...