Magnesiumchlorid Hexahydrat, EMPROVE® API JPC, USP, Ph. Eur., BP pharmaceutical_ingredient, SAFC®

Lieferant: Merck

Synonyme: Magnesium dichloride

1.02367.5000 1.02367.9029 1.02367.9525
1.02367.5000EA 343 EUR
1.02367.5000 1.02367.9029 1.02367.9525
Magnesiumchlorid Hexahydrat, EMPROVE® API JPC, USP, Ph. Eur., BP pharmaceutical_ingredient, SAFC®
Magnesiumchlorid Hexahydrat

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Standards: Corresponds to PhEur, BP, JPC, USP

Formel: MgCl₂·6H₂O
Molecular Weight: 203.3 g/mol
Siedepunkt: 1412
Schmelzpunkt: 117
Dichte: 1.57 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149781
CAS-Nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Bestellen

Spezifikation Testergebnisse

Assay (complexometric) 99.0 - 101.0 %
Identity passes test
Appearance of solution passes test
Insoluble matter ≤ 0.005 %
Acidity or alkalinity passes test
pH-value (5 %; water) 5.0 - 7.0
Bromide (Br) ≤ 0.05 %
Sulfate (SO₄) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.001 %
Al (Aluminium) ≤ 0.0001 %
As (Arsenic) ≤ 0.0002 %
Ba (Barium) passes test
Ca (Calcium) ≤ 0.01 %
Fe (Iron) ≤ 0.0005 %
Hg (Mercury) ≤ 0.0001 %
K (Potassium) ≤ 0.0500 %
NH₄ (Ammonium) ≤ 0.005 %
Pb (Lead) ≤ 0.0004 %
Water 51.0 - 55.0 %
Total aerobic microbial count ≤ 100 CFU/g
Total yeast and mould count ≤ 100 CFU/g
Endotoxins ≤ 3.0 I.U./g
Residual solvents (Ph Eur/USP/ICH) excluded by manufacturing process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR