Kaliumiodat, Certipur® volumetrischer Standard, sekundäres Referenzmaterial für Iodometrie traceable to NIST, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.02404.0100 1.02404.0103
1.02404.0100EA 169 EUR
1.02404.0100 1.02404.0103
Kaliumiodat, Certipur® volumetrischer Standard, sekundäres Referenzmaterial für Iodometrie traceable to NIST, Supelco®
Kaliumiodat

Merck have introduced into the range of Certipur® standards some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: KlO₃
Schmelzpunkt: 560 °C
Dichte: 3,98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011406
CAS-Nummer: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Bestellen

Spezifikation Testergebnisse

Solubility 92 g/l (25 °C)
Melting Point 560 °C decomposes
Molar Mass 214 g/mol
Bulk Density 2000 kg/m3
Density 3.98 g/cm3 (20 °C)
pH 6 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR