Ölrot O-Lösung Färbemittel für Histologie, für Zytologie, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: CI 26125

1.02419.0250
1.02419.0250EA 131 EUR
1.02419.0250
Ölrot O-Lösung Färbemittel für Histologie, für Zytologie, Sigma-Aldrich®
Ölrot O

Visualisation of neutral lipids in cryosections


This Oil red O solution is ready-to-use and results in a brilliant, bright red staining of lipds (triglycerides), while the nuclei are counterstained blue with Hematoxylin solution acc. to Gill II. Oil red O staining is used for the detection of neutral fat or polyethylene granula in e. g. histological tissues of human origin, in joint pathology or in cytological sample material. The stain is carried out on cryo sections of native tissue. Approx. 200 stains can be performed with one bottle of this IVD registered, CE certified staining solution.

Formel: C₂₆H₂₄N₄O
Molecular Weight: 408,51 g/mol
Dichte: 0,838 g/cm3
Flammpunkt: -11 °C
MDL: MFCD00003898
CAS-Nummer: 1320-06-5
UN: 1993
ADR: 3,II
IVDLogo

Bestellen

Spezifikation Testergebnisse

Suitability for microscopy passes test
Lipids bright red
Nuclei Blue

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR