Calciumchlorid Dihydrat, EMPROVE® API Ph. Eur., JP, USP, BP suitable for use as an active pharmaceutical ingredient, SAFC®

Lieferant: Merck
Achtung

Synonyme: Calcium Chloride Dihydrate, Calcium dichloride

1.02465.1000 1.02465.9029 1.02465.9525
1.02465.1000EA 192 EUR
1.02465.1000 1.02465.9029 1.02465.9525
Calciumchlorid Dihydrat, EMPROVE® API Ph. Eur., JP, USP, BP suitable for use as an active pharmaceutical ingredient, SAFC®
Calciumchlorid Dihydrat
Formel: CaCl₂·2H₂O
Molecular Weight: 147.01 g/mol
Siedepunkt: 1600
Schmelzpunkt: 176
Dichte: 1.85 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149613
CAS-Nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR