Calciumchlorid Dihydrat, EMPROVE® API Ph. Eur., JP, USP, BP suitable for use as an active pharmaceutical ingredient, SAFC®
Lieferant: Merck
|
Achtung
|
Synonyme:
Calcium Chloride Dihydrate, Calcium dichloride
1.02465.1000
1.02465.9029
1.02465.9525
1.02465.1000EA
192
EUR
1.02465.1000
1.02465.9029
1.02465.9525
Calciumchlorid Dihydrat, EMPROVE® API Ph. Eur., JP, USP, BP suitable for use as an active pharmaceutical ingredient, SAFC®
Calciumchlorid Dihydrat
Formel:
CaCl₂·2H₂O Molecular Weight: 147.01 g/mol Siedepunkt: 1600 Schmelzpunkt: 176 Dichte: 1.85 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149613 CAS-Nummer: 10035-04-8 EINECS: 233-140-8 Merck Index: 13,01660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...