Cobalt(II)sulfat Heptahydrat 97,0-102,0% (durch komplexometrische Analyse), reinst, SAFC®

Lieferant: Merck
Gefahr

Synonyme: Cobaltsulfat Heptahydrat

1.02546.9050
1.02546.9050EA 0 EUR
1.02546.9050
Cobalt(II)sulfat Heptahydrat 97,0-102,0% (durch komplexometrische Analyse), reinst, SAFC®
Cobalt(II)sulfat Heptahydrat
Formel: CoSO₄·7H₂O
Schmelzpunkt: 735 °C
Dichte: 2,03 g/cm³ (20 °C)
MDL: MFCD00149658
CAS-Nummer: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 12,02510

Bestellen

Spezifikation Testergebnisse

Assay (complexometric) 97.0 - 102.0 %
Identity conforms
Chloride (Cl) ≤ 0.02 %
Fe (Iron) ≤ 0.005 %
Pb (Lead) ≤ 0.005 %
Residual solvents (according to ICH-guideline) excluded by manufacturing process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR