Ammoniumnitrat-Lösung 1 mol/l acc. to DIN 19730, Supelco®
Lieferant: Merck
Synonyme:
Ammonsalpeter
M-Clarity™ Qualitätsstufe = MQ100
1.02596.2500
1.02596.2500EA
389
EUR
1.02596.2500
Ammoniumnitrat-Lösung 1 mol/l acc. to DIN 19730, Supelco®
Ammoniumnitrat
Formel:
NH₄NO₃ Molecular Weight: 80,04 g/mol |
MDL:
MFCD00011425 CAS-Nummer: 6484-52-2 |
Spezifikation Testergebnisse
Solubility | Soluble at 20 °C |
Boiling Point | 100 °C (1013 hPa) |
Density | 1.03 g/cm3 (20 °C) |
Storage temeprature | +15...+25 °C |
Assay | 0,99 - 1,01 mol/l |
pH-value | 5 - 6 |
As (Arsenic) | ≤0,005 ppm |
Be (Beryllium) | ≤0,001 ppm |
Cd (Cadmium) | ≤0,002 ppm |
Co (Cobalt) | ≤0,005 ppm |
Cr (Chromium) | ≤0,005 ppm |
Cu (Copper) | ≤0,005 ppm |
Hg (Mercury) | ≤0,001 ppm |
Mg (Magnesium) | ≤0,001 ppm |
Mo (Molybdenum) | ≤0,005 ppm |
Ni (Nickel) | ≤0,010 ppm |
Pb (Lead) | ≤0,005 ppm |
Sb (Antimony) | ≤0,005 ppm |
Tl (Thallium) | ≤0,005 ppm |
U (Uranium) | ≤0,001 ppm |
V (Vanadium) | ≤0,005 ppm |
Zn (Zinc) | ≤0,005 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...