Kupfer(II)sulfat Pentahydrat, EMSURE® ISO, ACS, Reag. Ph. Eur. analysis_for, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Kupfersulfat Pentahydrat, Kupfersulfat-Pentahydrat
MQ300
1.02790.0250
1.02790.1000
1.02790.5000
1.02790.9050
1.02790.0250EA
48
EUR
1.02790.0250
1.02790.1000
1.02790.5000
1.02790.9050
Kupfer(II)sulfat Pentahydrat, EMSURE® ISO, ACS, Reag. Ph. Eur. analysis_for, Supelco®
Kupfer(II)sulfat Pentahydrat
Formel:
CuSO₄·5H₂O Molecular Weight: 249.69 g/mol Dichte: 2.284 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149681 CAS-Nummer: 7758-99-8 EINECS: 231-847-6 UN: 3077 ADR: 9 III REACH: 01-2119520566-40 Merck Index: 13,02682 |
Spezifikation Testergebnisse
Assay (iodometric) | 99.0-100.5% |
Insoluble matter | 0.005% max |
Chloride (Cl) | 0.0005% max |
Total nitrogen (N) | 0.001% max |
Calcium (Ca) | 0.005% max |
Iron (Fe) | 0.003% max |
Potassium (K) | 0.001% max |
Sodium(Na) | 0.005% max |
Nickel (Ni) | 0.005% max |
Lead (Pb) | 0.005% max |
Zinc (Zn) | 0.03% max |
Corresponds to ACS,ISO,Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...