Cyclohexan, SupraSolv® für die Gaschromatographie, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

1.02817.1000 1.02817.2500 1.02817.4000
1.02817.1000EA 114 EUR
1.02817.1000 1.02817.2500 1.02817.4000
Cyclohexan, SupraSolv® für die Gaschromatographie, Supelco®
Cyclohexan

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionisation detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formel: C₆H₁₂
Siedepunkt: 81 °C (1013 hPa)
Schmelzpunkt: 7 °C
Dichte: 0,7785 g/cm³ (20 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solubility 55 mg/l (20 °C)
Melting Point 6 °C
Molar Mass 84.16 g/mol
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260 °C
Viscosity kinematic 1.26 mm2/s (20 °C)
Evaporation number 3.5
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR