Cyclohexan, EMPLURA®, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

1.02832.1000 1.02832.2500 1.02832.6025 1.02832.6190 1.02832.9190
1.02832.1000EA 56.2 EUR
1.02832.1000 1.02832.2500 1.02832.6025 1.02832.6190 1.02832.9190
Cyclohexan, EMPLURA®, Supelco®
Cyclohexan

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formel: C₆H₁₂
Siedepunkt: 81 °C (1013 hPa)
Schmelzpunkt: 7 °C
Dichte: 0,7785 g/cm³ (20 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR