D(-)-Sorbit F, Liquid E420, SAFC®

Lieferant: Merck

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

1.02993.9903
1.02993.9903EA 0 EUR
1.02993.9903
D(-)-Sorbit F, Liquid E420, SAFC®
D(-)-Sorbit

Sorbitol F liquid (sorbitol syrup, noncrystallizing)

Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Dichte: 1.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR