D(-)-Sorbit 70% 70, Liquid, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, E420 suitable for use as excipient, SAFC®

Lieferant: Merck

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

1.02994.9050 1.02994.9270
1.02994.9050EA 672 EUR
1.02994.9050 1.02994.9270
D(-)-Sorbit 70% 70, Liquid, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, E420 suitable for use as excipient, SAFC®
D(-)-Sorbit

Sorbitol flüssig (Sorbitsirup, kristallisierend)
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (#).
Corresponds to Ph. Eur., BP, USP, E 420.
Conforms to the purity criteria on food additives according to the current European Commission Regulation

Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Dichte: 1.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Solid matter 69.0 - 72.0 %
Sorbitol (HPLC, calc.on anhydrous substance) 92.0 - 101.0 %
Sorbitol (HPLC) ≥ 65.0 %
Identity passes test
Appearance of solution (10 %; water) passes test
pH-value (14 %; water) 5.0 - 7.0
conductivity (25°C) ≤ 10
As (Arsenic) ≤ 0.0001 %
Ni (Nickel) (#) ≤ 0.0001 %
Pb (Lead) ≤ 0.00005 %
Ethylene glycol (HPLC) ≤ 0.10 %
Diethylene glycol (HPLC) ≤ 0.10 %
Reducing sugars after hydrolysis/total sugar (as glucose) ≤ 0.5
Reducing sugars (as glucose) ≤ 0.2
Sulfated ash ≤ 0.1
Water 28.5 - 31.0 %
Residues of solvents (ICH Q3C) excluded by manufacturing process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR