Bromthymolblau ACS, Reag. Ph. Eur.
Lieferant: Merck
Synonyme:
3′,3′′-Dibromthymolsulfonphthalein, BTB
M-Clarity™ Qualitätsstufe = MQ200
1.03026.0025
1.03026.0500
1.03026.0025EA
180
EUR
1.03026.0025
1.03026.0500
Bromthymolblau ACS, Reag. Ph. Eur.
Bromthymolblau
Formel:
C₂₇H₂₈Br₂O₅S Schmelzpunkt: 204…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005872 CAS-Nummer: 76-59-5 EINECS: 200-971-2 Merck Index: 13,01430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...