2,6-Dichlorchinon-4-chlorimid, GR Reag. Ph. Eur., Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
1.03037.0010
1.03037.0100
1.03037.0010EA
515
EUR
1.03037.0010
1.03037.0100
2,6-Dichlorchinon-4-chlorimid, GR Reag. Ph. Eur., Supelco®
2,6-Dichlorchinon-4-chlorimid
Formel:
C₆H₂Cl₃NO Lagertemperatur: Kühlschrank |
MDL:
MFCD00001611 CAS-Nummer: 101-38-2 EINECS: 202-937-2 UN: 3224 ADR: 4.1, |
Spezifikation Testergebnisse
Assay (iodometric) | ≥ 98 |
Identity (IR-spectrum) | passes test |
Spec. Absorptivity A 1%/1cm (312 nm; 0,001 %; 1 cm; ethanol) | ≥ 800 |
Melting point | 65 - 68 |
Hazard Class | 4.1 |
Hazard Class Description | Flammable solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...