1,5-Diphenylcarbazon (cont. 50 % Diphenylcarbazide) ACS, Reag. Ph. Eur., Supelco®

Lieferant: Merck

Synonyme: sym-Diphenylcarbazone, Phenylazoameisensäure-2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazide

M-Clarity™ Qualitätsstufe = MQ100

1.03087.0025
1.03087.0025EA 400 EUR
1.03087.0025
1,5-Diphenylcarbazon (cont. 50 % Diphenylcarbazide) ACS, Reag. Ph. Eur., Supelco®
1,5-Diphenylcarbazon
Formel: C₁₃H₁₂N₄O
Molecular Weight: 240,26 g/mol
MDL: MFCD00003024
CAS-Nummer: 538-62-5

Bestellen

Spezifikation Testergebnisse

Solubility passes test
Melting Point 153 - 158 °C (decomposition)
Bulk Density 380 kg/m3
Bulk density 380 kg/m3
Storage +15...+25 °C
Purity 34,5 - 40,4 % (a/a)
Identity passes test
Substances insoluble in ethanol passes test
Melting point 152,5 - 158,4 °C
Sulfated ash ≤0,1 %
Suitability for mercury determination passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR