Diethanolamin-Lösung 85% zur Synthese (max. 0.5% ethanolamine)

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.03109.5000
1.03109.5000EA 0 EUR
1.03109.5000
Diethanolamin-Lösung 85% zur Synthese (max. 0.5% ethanolamine)
Diethanolamin
Formel: C₄H₁₁NO₂
MDL: MFCD00002843
CAS-Nummer: 111-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR