Ammoniumeisen(III)citrat (∼18% Fe), SAFC®

Lieferant: Merck

Synonyme: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

1.03762.1000
1.03762.1000EA 319 EUR
1.03762.1000
Ammoniumeisen(III)citrat (∼18% Fe), SAFC®
Ammoniumeisen(III)citrat
Formel: C₆H₅+4y)Fe(x)N(y)O₇
MDL: MFCD00013099
CAS-Nummer: 1185-57-5
EINECS: 214-686-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR