Glycerin 98,0-101,0% (durch Alkalimetrie) Reag. Ph. Eur., ACS
Lieferant: Merck
Synonyme:
1,2,3-Propantriol, Glycerol
M-Clarity™ Qualitätsstufe = MQ300
1.04057.9011
1.04057.2511
1.04057.9026
1.04057.9011EA
1060
EUR
1.04057.9011
1.04057.2511
1.04057.9026
Glycerin 98,0-101,0% (durch Alkalimetrie) Reag. Ph. Eur., ACS
Glycerin
Bio-Based Glycerol
Our bio-based glycerol is produced from rapeseed, a renewable raw material, and a by-product of biodiesel production. It offers numerous advantages compared to petroleum-derived glycerol.
- Non-toxic, nonirritating
- Nonflammable (flash point 160 °C, autoflammability 400 °C)
- No waste due to 100% biodegradability
- Low vapor pressure (environmental advantage)
- Reliable availability (independent of petrochemical production)
Formel:
HOCH₂CH(OH)CH₂OH Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 18…20 °C Dichte: 1,26 g/cm³ (20 °C) Flammpunkt: 199 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 |
|
Spezifikation Testergebnisse
Assay (GC, Calculated on Anhydrous Substance) | ≤99.5 % |
Assay (Alkalimetric, Calculated on Anhydrous Substance) | 98.0 - 101.0% |
Identity | Passes Test |
Colour | ≤10 APHA |
Acidity or Alkalinity | Passes Test |
Free Acid (CH₃COOH) | ≤0.002 % |
Neutrality | Passes Test |
Refractive Index (n 20/D) | 1.470 - 1.475 |
Chloride (Cl) | ≤0.0010% |
Sulphate (SO₄) | ≤0.001% |
Halogenated Compounds (as Cl) | ≤0.0030% |
Heavy Metals (as Pb) | ≤0.00020% |
Al (Aluminium) | ≤0.00005% |
B (Boron) | ≤0.000002% |
Ba (Barium) | ≤0.00001% |
Ca (Calcium) | ≤0.0001% |
Cd (Cadmium) | ≤0.000005% |
Co (Cobalt) | ≤0.000005% |
Cr (Chromium) | ≤0.000002% |
Cu (Copper) | ≤0.000005% |
Fe (Iron) | ≤0.00001% |
Mg (Magnesium) | ≤0.0001% |
Mn (Manganese) | ≤0.00001% |
Ni (Nickel) | ≤0.000002% |
Pb (Lead) | ≤0.00001% |
Sn (Tin) | ≤0.00001% |
Zn (Zinc) | ≤0.00001% |
Methanol | ≤3000 ppm |
Other Residual Solvents (ICH Q3C) | Excluded by Production Process |
Acrolein and Glucose | Passes Test |
Aldehydes | ≤10 ppm |
Diethylene Glycol and Related Substances (Impurity A) | ≤0.1% |
Diethylene Glycol and Related Substances (Any Further Impurity) | ≤0.1% |
Diethylene Glycol and Related Substances (Sum of All Impurities) | ≤0.5% |
Fatty Acids and Esters (as Butyric Acid) | ≤0.05% |
Substances Darkened by Sulphuric Acid | Passes Test |
Sugar | Passes Test |
Sulfated Ash | ≤0.005% |
Water | ≤0.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...