D(+)-Glucose Monohydrat, Millipore®
Lieferant: Merck
Synonyme:
Dextrose Monohydrat, Traubenzucker Monohydrat
M-Clarity™ Qualitätsstufe = MQ100
1.04074.0500
1.04074.1000
1.04074.5000
1.04074.9050
1.04074.0500EA
36.2
EUR
1.04074.0500
1.04074.1000
1.04074.5000
1.04074.9050
D(+)-Glucose Monohydrat, Millipore®
D(+)-Glucose Monohydrat
Formel:
C₆H₁₂O₆·H₂O Schmelzpunkt: 83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149450 CAS-Nummer: 14431-43-7 EINECS: 200-075-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...