Glyoxalbis(2-hydroxyanil), GR zur Analyse, Supelco®
Lieferant: Merck
Synonyme:
2,2'-Ethanediylidenedinitrilodiphenol, 2,2'-(Ethane-1,2-diylidenebis(azanylylidene))diphenol, Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane
M-Clarity™ Qualitätsstufe = MQ100
1.04191.0025
1.04191.0025EA
295
EUR
1.04191.0025
Glyoxalbis(2-hydroxyanil), GR zur Analyse, Supelco®
Glyoxalbis(2-hydroxyanil)
Formel:
C₁₄H₁₂N₂O₂ Molecular Weight: 240,26 g/mol |
MDL:
MFCD00002183 CAS-Nummer: 1149-16-2 |
Spezifikation Testergebnisse
Assay (ex N) | ≥97 |
Identity (IR-spectrum) | Passes test |
Melting point | about 200°C |
Sulfated ash | ≤0,1% |
Loss on Drying (105°C) | ≤ 0.5 % |
Suitability for metal determination | Passes test |
Solubility | (20 °C) slightly soluble |
Storage | +15...+25 °C |
Loss on drying (105 °C) | ≤0,5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...