Graphit, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

1.04206.9025
1.04206.9025EA 0 EUR
1.04206.9025
Graphit, Supelco®
Graphit
Formel: C
Molecular Weight: 12,01 g/mol
Schmelzpunkt: 3827 °C
Dichte: 2,09…2.23 g/cm³ (20 °C)
MDL: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR